Merck
The uses and properties of almond oil.
Complementary therapies in clinical practice (2010-02-05)
Zeeshan Ahmad
ABSTRACT
Almond oil [Oleum amygdalae] has long been used in complementary medicine circles for its numerous health benefits. Although no conclusive scientific data exists currently, almonds and almond oil have many properties including anti-inflammatory, immunity-boosting and anti-hepatotoxicity effects. Further, associations between almond oil and improved bowel transit have been made, which consequently reduces irritable bowel syndrome symptoms. Further, some studies show a reduced incidence of colonic cancer. Moreover, cardiovascular benefits have also been identified with almond oil elevating the levels of so-called 'good cholesterol', high-density lipoproteins (HDL), whilst it reduces low-density lipoproteins (LDL). Historically, almond oil had been used in Ancient Chinese, Ayurvedic and Greco-Persian schools of Medicine to treat dry skin conditions such as psoriasis and eczema. Further, it is through anecdotal evidence and clinical experiences that almond oil seemingly reduces hypertrophic scarring post-operatively, smoothes and rejuvenates skin. Almond oil has emollient and sclerosant properties and, therefore, has been used to improve complexion and skin tone. Further studies looking into the use of almond oil post-operatively for the reduction of scarring are suggested.