Merck
  • In vivo hepatic oxidative stress because of carbon tetrachloride toxicity: protection by melatonin and pinoline.
In vivo hepatic oxidative stress because of carbon tetrachloride toxicity: protection by melatonin and pinoline.
Journal of pineal research (2010-06-08)
M Aranda, C D Albendea, F Lostalé, L López-Pingarrón, L Fuentes-Broto, E Martínez-Ballarín, R J Reiter, M C Pérez-Castejón, J J García
ABSTRACT
The protective in vivo effects of melatonin or pinoline on carbon tetrachloride (CCl(4))-induced oxidative damage were investigated in liver of rats and compared to rats injected only with CCl(4) (5 mL/kg body weight). Hepatic cell membrane fluidity, monitored using fluorescence spectroscopy, exhibited a significant decrease in animals exposed to CCl(4) compared to control rats. Increases in lipid and protein oxidation, as assessed by concentrations of malondialdehyde (MDA) and 4-hydroxyalkenals (4-HDA), and protein carbonylation, respectively, were also seen in hepatic homogenates of animals exposed to CCl(4). The administration of melatonin (10 mg/kg body weight) or pinoline injected 30 min before and 1 hr after CCl(4), fully prevented membrane rigidity and protein oxidation. However, treatment with melatonin was more effective in terms of reducing lipid peroxidation than pinoline, as the increases in MDA+4-HDA levels because of CCl(4) were reduced by 93.4% and 34.4% for melatonin or pinoline, respectively. Livers from CCl(4)-injected rats showed several histopathological alterations; above all, there were signs of necrosis and ballooning degeneration. The concurrent administration of melatonin or pinoline reduced the severity of these morphological changes. On the basis of the biochemical and histopathological findings, we conclude that both melatonin and pinoline were highly effective in protecting the liver against oxidative damage and membrane rigidity because of CCl(4). Therefore, these indoles may be useful as cotreatments for patients with hepatic intoxication induced by CCl(4).