Merck
  • A preliminary study on the use of meconium for the assessment of prenatal exposure to heavy metals in Japan.
A preliminary study on the use of meconium for the assessment of prenatal exposure to heavy metals in Japan.
Journal of UOEH (2013-06-19)
Yeonsil Yang, Satoshi Nakai, Shiori Oda, Hiroko Nishino, Misato Ishii, Hiroko Yokoyama, Hideaki Matsuki
ABSTRACT
In recent years, there has been increasing interest in the relevance of environmental pollution to children's health. We investigated whether the concentrations of heavy metals in meconium can be used for the assessmet of prenatal exposure in Japan. Our study subjects were newborns whose mothers were hospitalized for childbirth at Tokai University Hospital between November 2010 and March 2011. The target compounds of this study were lead (Pb), cadmium (Cd) and arsenic (As). Meconium samples collected from 102 newborns were analyzed using ICPMS. The median concentrations of Pb and As in the meconium were 0.71 ppb and 0.03 ppb, respectively. The median concentration of Cd was below the limit of quantification. These concentration values are much lower than those used in previous studies. Further study is needed to compare the concentrations of heavy metals in biological samples and to investigate the availability of meconium for the assessment of exposure to heavy metals during pregnancy.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Arsenic, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Arsenic, powder, ≥99.997% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis