Merck
  • Investigation of transport of genistein, daidzein and their inclusion complexes prepared with different cyclodextrins on Caco-2 cell line.
Investigation of transport of genistein, daidzein and their inclusion complexes prepared with different cyclodextrins on Caco-2 cell line.
Journal of pharmaceutical and biomedical analysis (2013-07-03)
Agnes Emma Daruházi, Tímea Kiss, Miklós Vecsernyés, Lajos Szente, Eva Szőke, Eva Lemberkovics
ABSTRACT
Isoflavonoids are widespread constituents in medical plants especially in legumes (Fabaceae), but occur in other different plant families as well (Rosaceae, Iridaceae, Amaranthaceae). Their antioxidant, estrogen-like, anti-inflammatory and analgesic effects make them promising compounds in therapy of important disorders especially in estrogen related diseases. Poor solubility in aqueous system of genistein and daidzein needs a solubility enhancement for pharmaceutical use. These compounds are suitable guest molecules for inclusion complex formation with cyclodextrins (CDs) considering matching their size and polarity. The molecular encapsulation with beta-cyclodextrin (β-CD), gamma-cyclodextrin (γ-CD), hydroxypropyl-beta-cyclodextrin (HP-β-CD) and random methyl-beta cyclodextrin (RAMEB-CD) results in a solid, molecularly dispersed form and in a significantly improved aqueous solubility of genistein and daidzein. Determining enhancement in solubility and bioavailability we investigated the transport of these inclusion complexes across Caco-2 cell line comparing that of the pure compounds and found significant improving effect of the different CD derivatives on membrane permeation of the two isoflavone aglycons.
Product Description
Supelco
Genistein, analytical standard
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Supelco
Daidzein, analytical standard