Merck
  • Helicobacter pylori-induced loss of survivin and gastric cell viability is attributable to secreted bacterial gamma-glutamyl transpeptidase activity.
Helicobacter pylori-induced loss of survivin and gastric cell viability is attributable to secreted bacterial gamma-glutamyl transpeptidase activity.
The Journal of infectious diseases (2013-07-13)
Manuel Valenzuela, Denisse Bravo, Jimena Canales, Carlos Sanhueza, Natalia Díaz, Oscar Almarza, Héctor Toledo, Andrew F G Quest
ABSTRACT
Helicobacter pylori is the etiologic agent of a series of gastric pathologies that may culminate in the development of gastric adenocarcinoma. An initial step in this process is the loss of glandular structures in the gastric mucosa, presumably as the consequence of increased apoptosis and reduced cellular regeneration, which may be attributed to the combination of several bacterial and host factors and to an unfavorable proinflammatory environment. In a previous study, we showed that survivin, a member of the inhibitor of apoptosis protein family, is expressed in the normal human gastric mucosa and that its levels decrease in the mucosa of infected patients and in gastric cells exposed in culture to the bacteria, coincident with increased cell death in the latter case. We investigated the bacterial factors responsible for loss of survivin in gastric cells exposed to H. pylori. The results of this study indicated that the loss of survivin due to H. pylori infection involves proteasome-mediated degradation of the protein. Studies with isogenic mutants deficient in either CagA, VacA, lipopolysaccharide, or gamma-glutamyl transpeptidase (GGT) implicated the latter in H. pylori-induced loss of survivin and cell viability. Moreover, experiments with the GGT inhibitor 6-diazo-5-oxo-l-norleucine and purified recombinant GGT protein indicated that secreted bacterial GGT activity was required and sufficient to induce these effects.
Product Description
γ-Glutamyl Transferase, ERM®, certified reference material
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid