Merck
  • Near-infrared pigments based on ion-pair charge transfer salts of dicationic and dianionic metal-dithiolene [M(II) = Pd, Pt] complexes.
Near-infrared pigments based on ion-pair charge transfer salts of dicationic and dianionic metal-dithiolene [M(II) = Pd, Pt] complexes.
Dalton transactions (Cambridge, England : 2003) (2013-07-19)
Davide Espa, Luca Pilia, Luciano Marchiò, Maria Laura Mercuri, Angela Serpe, Elisa Sessini, Paola Deplano
ABSTRACT
Mixing [M(Et2dazdt)2](BF4)2 [M = Ni(II), Pd(II), Pt(II); Et2dazdt = N,N'-diethyl-perhydrodiazepine-2,3-dithione] with (Bu4N)2[M(mnt)2] (mnt = maleonitrile-2,3-dithiolate) in CH3CN produces the known mixed ligand dithiolene complex [Ni(Et2dazdt)(mnt)] in the nickel case and ion-pair salts [M(Et2dazdt)2][M(mnt)2] in the palladium (1) and platinum (2) cases. Structural characterization of 2 shows that the anionic (donor) and cationic (acceptor) complexes form an irregular stack that lies in the bc crystallographic plane. The shortest contacts exchanged by the anion and cation molecules within each stack are those occurring through the hydrogen atoms of the CH2 groups of Et2dazdt and the Pt(2)-S(22) segment (d(H(81a)-S(22) = 2.981(3) Å) and the nitrogen atom of the cyano group of mnt and the carbon atom of one of the thione moieties (d(N(12)-C(11) = 3.179(3) Å). The Pt atom of [Pt(mnt)2](2-) is surrounded by two hydrogen atoms of the Et2dazdt ligand, whereas the Pt atom of [Pt(Et2dazdt)2](2+) is surrounded by two carbon atoms of the dithiolate moiety of mnt. Intramolecular interactions are due to contacts exchanged mainly through H-atoms, which are suitable to mediate charge-transfer (CT) interactions. In fact, these salts are characterized by a long wavelength CT peak [λmax = 905 nm (1), 937 nm (2)], which makes them candidates as near-infrared pigments, whose properties are tunable with the redox features of the components, the energy of the NIR absorption being relatable to the driving force of electron transfer from the donor (dianion) to the acceptor (dication). A thorough description of interactions occurring between the complex anions and complex cations has been achieved by investigating the Hirshfeld surface (HS) properties. Computational methods are in agreement with experimental findings and allow us to highlight the electronic features of the components of these CT salts, providing a structure-property relationship, useful in designing new candidates to optimize the desired properties.
Sigma-Aldrich
Toluene-3,4-dithiol, technical grade, 90%
Supelco
Toluene-3,4-dithiol, for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Toluene, analytical standard
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol