Merck
Mineralized cyclodextrin nanoparticles for sustained protein delivery.
Carbohydrate polymers (2013-08-06)
Maharajan Sivasubramanian, Thavasyappan Thambi, Jae Hyung Park
ABSTRACT
The extensive therapeutic potential of protein drugs has been severely limited by their instability and short biological half-lives in vivo. To prolong their therapeutic effects, a sustained delivery system is required. In this study, cyclodextrin-based polymeric nanoparticles (CD-NPs), mineralized by calcium phosphate as the diffusion barrier, were developed as a carrier for sustained protein delivery. Spherical CD-NPs were readily prepared by a conjugate, composed of β-CD as the protein-binding moiety and carboxymethyl dextran as the substrate for mineralization in a physiological solution. Owing to the presence of carboxylic acids in CD-NPs, they were effectively mineralized by sequential addition of calcium nitrate and ammonium phosphate. The physicochemical characteristics of mineralized CD-NPs were characterized using FT-IR, thermogravimetric analysis, transmission electron microscopy and energy dispersive X-ray photoelectron spectroscopy. Mineralization reduced CD-NP particle size from 310 nm to 121 nm in PBS (pH 7.4) indicating the formation of compact nanoparticles. Carbonic anhydrase B (CAB), chosen as the model protein, was loaded into the mineralized CD-NPs with a high loading efficiency (80%) by a simple dialysis method. In vitro release tests showed that CAB was completely released from bare CD-NPs in 3 days. Interestingly, the mineralized CD-NPs released CAB in a sustained manner for 21 days, which was due to the stable calcium phosphate barrier inhibiting CAB release. The enzymatic activity of CAB, which was released from the nanoparticles, did not significantly deteriorate compared to native CAB. Overall, mineralized CD-NPs could be a promising carrier for sustained protein delivery.
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 25,000
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 50,000
Supelco
Dextran, analytical standard, 25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran, analytical standard, 12,000
Supelco
Dextran, analytical standard, 410,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 5,000
Supelco
Dextran, analytical standard, 1,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000