Merck
  • Effect of temperature gradients on single-strand conformation polymorphism analysis in a capillary electrophoresis system using Pluronic polymer matrix.
Effect of temperature gradients on single-strand conformation polymorphism analysis in a capillary electrophoresis system using Pluronic polymer matrix.
Analytica chimica acta (2013-08-21)
Hee Sung Hwang, Gi Won Shin, Han Jin Park, Chang Y Ryu, Gyoo Yeol Jung
ABSTRACT
Capillary electrophoresis-single strand conformation polymorphism (CE-SSCP) analysis is a prominent bioseparation method based on the mobility diversity caused by sequence-induced conformational differences of single-stranded DNA. The use of Pluronic polymer matrix has opened up new opportunities for CE-SSCP, because it improved the resolution for various genetic analyses. However, there still exists a challenge in optimizing Pluronic-based CE-SSCP, because the physical properties of Pluronic solutions are sensitive to temperature, particularly near the gelation temperature, where the viscoelasticity of Pluronic F108 solutions sharply changes from that of a Newtonian fluid to a hydrogel upon heating. We have focused on a set of experiments to control the ambient temperature of the CE system with the aim of enhancing the reliability of the CE-SSCP analysis by using the Applied Biosystems ABI 3130xl genetic analyzer with Pluronic F108 solution matrix. The ambient temperature control allowed us to vary the inlet and outlet portion of the capillary column, while the temperature of the column was kept at 35°C. The resolution to separate 2 single-base-pair-differing DNA fragments was significantly enhanced by changing the temperature from 19 to 30°C. The viscoelastic properties of the F108 solution matrix upon heating were also investigated by ex situ rheological experiments with an effort to reveal how the development of gels in Pluronic solutions affects the resolution of CE-SSCP. We found that the column inlet and outlet temperatures of the capillary column have to be controlled to optimize the resolution in CE-SSCP by using the Pluronic matrix.
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil