Merck
  • Partly reversible central auditory dysfunction induced by cerebral vasospasm after subarachnoid hemorrhage.
Partly reversible central auditory dysfunction induced by cerebral vasospasm after subarachnoid hemorrhage.
Journal of neurosurgery (2013-08-27)
Ester Ponzetto, Marco Vinetti, Cécile Grandin, Thierry Duprez, Vincent van Pesch, Naïma Deggouj, Renaud Lhommel, Philippe Hantson
ABSTRACT
The authors describe a rare case of central auditory dysfunction induced by cerebral vasospasm after aneurysmal subarachnoid hemorrhage (SAH). A 55-year-old woman who was admitted after aneurysmal SAH developed cerebral vasospasm on Day 3 affecting mainly the right middle cerebral artery (MCA) and partly the left MCA. The vasospasm became refractory to conventional therapy and was ultimately improved by intraarterial infusion of nimodipine in the right MCA and angioplasty. Severe auditory dysfunction was apparent from Day 10 as the patient was not reactive to speech or environmental sounds. Brain MRI on Day 17 demonstrated infarcted areas mainly in the right hippocampus, medial occipital lobe, and thalamus. The patient underwent further examination using audiometry, speech testing, auditory evoked potentials, functional MRI, and cerebral PET. The initial diagnosis was extended nonverbal agnosia and total pure word deafness. The central auditory dysfunction improved over 6 months, with persisting hyperacusis, tinnitus, and amusia. Central auditory dysfunction is a rare complication after SAH. While cortical deafness may be associated with bilateral lesions of the temporal cortex, partly reversible central auditory dysfunction was observed in this patient after prominently unilateral right temporal lesions. The role of the interthalamic connections can be discussed, as well as the possibility that a less severe vasospasm on the left MCA could have transiently impaired the left thalamocortical auditory pathways.
Sigma-Aldrich
Nimodipine
Pricing and availability is not currently available.