Merck
  • Distinct tRNA modifications in the thermo-acidophilic archaeon, Thermoplasma acidophilum.
Distinct tRNA modifications in the thermo-acidophilic archaeon, Thermoplasma acidophilum.
FEBS letters (2013-10-01)
Chie Tomikawa, Takayuki Ohira, Yasushi Inoue, Takuya Kawamura, Akihiko Yamagishi, Tsutomu Suzuki, Hiroyuki Hori
ABSTRACT
Thermoplasma acidophilum is a thermo-acidophilic archaeon. We purified tRNA(Leu) (UAG) from T. acidophilum using a solid-phase DNA probe method and determined the RNA sequence after determining via nucleoside analysis and m(7)G-specific aniline cleavage because it has been reported that T. acidophilum tRNA contains m(7)G, which is generally not found in archaeal tRNAs. RNA sequencing and liquid chromatography-mass spectrometry revealed that the m(7)G modification exists at a novel position 49. Furthermore, we found several distinct modifications, which have not previously been found in archaeal tRNA, such as 4-thiouridine9, archaeosine13 and 5-carbamoylmethyuridine34. The related tRNA modification enzymes and their genes are discussed.
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
Guanosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanosine, ≥97.0% (HPLC)