Merck
  • Construction of a supF-based system for detection of mutations in the chromosomal DNA of Arabidopsis.
Construction of a supF-based system for detection of mutations in the chromosomal DNA of Arabidopsis.
Molecular genetics and genomics : MGG (2013-10-25)
Keiichiro Hiratsu, Shiori Shiotani, Kozo Makino, Tatsuo Nunoshiba
ABSTRACT
The factors maintaining genomic integrity, which have been studied in detail in other species, have yet to be investigated in plants. Recent progress in gene-silencing technology has made it possible to produce transgenic plants with loss-of-function phenotypes for the effective analysis of these factors, even with the high redundancy of genes in plants. Therefore, a mutation-detection system for plants is necessary to estimate the biological function of a target gene for mutation frequencies and spectra. Here, we reported the development of a novel system to analyze mutations in the chromosomal DNA of plants. The supF gene of E. coli was used as a target for the mutation because it was possible to detect all mutational base changes. Based on the plasmid pTN30, which carries supF, we constructed a binary Ti vector for its introduction to Arabidopsis genomes. The system was validated by measuring mutations in both non-treated and mutagen-treated transgenic plants. DNA fragments including pTN30 were rescued from the plants, and introduced into E. coli KS40/pOF105 to isolate the supF mutant clones conferring both nalidixic acid and streptomycin resistance on transformants. We found that the mutation frequency was approximately three times higher with the ethyl methanesulfonate (EMS) treatment than without it and G:C to A:T transitions dominated, which was the most reasonable mutation induced by EMS. These results show that this system allowed for the rapid analysis of mutations in plants, and may be useful for analyzing plant genes related to the functions of genomic stability and monitoring environmental genotoxic substances.
Product Description
Supelco
Ethyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
Ethyl methanesulfonate, liquid
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution