Merck
  • The effect of numbers and locations of retentive holes placed on master casts on reducing the polymerization distortion of the maxillary complete denture.
The effect of numbers and locations of retentive holes placed on master casts on reducing the polymerization distortion of the maxillary complete denture.
The European journal of prosthodontics and restorative dentistry (2013-11-23)
M Golshah, M S Monfared, S Najafi-Abrandabadi
ABSTRACT
Polymerization shrinkage is one of the disadvantages ofpolymethyl methacrylate resulting in distortion of resin denture bases. The aim of this study is to analyze the effect of numbers and locations of retentive holes on reducing polymerization distortion. Forty eight master casts were assigned as: control group (A); one anchoring hole (B); three anchoring holes (C); five anchoring holes (D).The gap distances between the denture bases and casts were measured. The data were analyzed with one-way analysis of variance (ANOVA) followed by the Post-Hoc LSD test. Mechanical anchorage reduced the gap distances in lateral and mid-palate significantly; and this decrease was affected by numbers and locations of anchoring holes significantly.
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000