Merck
Treatment of pediatric anxiety disorders.
Annals of the New York Academy of Sciences (2013-11-28)
Amy Rapp, Alice Dodds, John T Walkup, Moira Rynn
ABSTRACT
This article provides a brief review of the current available data concerning present treatment and potential new treatment advances for pediatric anxiety disorders, such as generalized anxiety disorder, separation anxiety disorder, social phobia, obsessive-compulsive disorder, and posttraumatic stress disorder. Disorder-specific treatment methods and innovations, particularly computer-assisted methods of delivery for cognitive behavioral therapy (CBT) will be reviewed. Additionally, the paper will discuss novel psychopharmacological compounds (e.g., D-cycloserine, riluzole, memantine, and anticonvulsant medications). Available evidence for the efficacy of novel medication strategies in adult studies and implications for their use in pediatrics will be discussed.
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
D-Cycloserine, synthetic
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
Riluzole, solid