Merck
  • Fluoride promotes osteoblastic differentiation through canonical Wnt/β-catenin signaling pathway.
Fluoride promotes osteoblastic differentiation through canonical Wnt/β-catenin signaling pathway.
Toxicology letters (2013-12-05)
Leilei Pan, Xiaoguang Shi, Shuang Liu, Xiaoying Guo, Ming Zhao, Ruoxin Cai, Guifan Sun
ABSTRACT
Although fluoride is known to stimulate bone formation, the underlying mechanisms are not fully understood. Recent studies have implicated the Wnt/β-catenin pathway as a major signaling cascade in bone biology. Our earlier studies highlighted a probable role of canonical Wnt pathway in bone formation of chronic fluoride-exposed rats, but the mechanism remains unclear. The current study determined the involvement of Wnt/β-catenin signaling in fluoride-induced osteoblastic differentiation. Using primary rat osteoblasts, we demonstrated that fluoride significantly promoted osteoblasts proliferation and alkaline phosphate (ALP) expression as well as the mRNA expression levels of bone differentiation markers, including type I collagen (COL1A1), ALP and osteonectin. We further found fluoride induced phosphorylations at serine 473 of Akt and serine 9 of glycogen synthase kinase-3β (GSK3β), which resulted in GSK-3β inhibition and subsequently the nuclear accumulation of the β-catenin, as shown by Western blot and immunofluorescence analysis. Moreover, fluoride also induced the expression of Wnt-targeted gene runt-related transcription factor 2 (Runx2). Importantly, the positive effect of fluoride on ALP activity and mRNA expressions of COL1A1, ALP, osteonection and Runx2 was abolished by DKK-1, a blocker of the Wnt/β-catenin receptor. Taken together, these findings suggest that fluoride promotes osteoblastic differentiation through Akt- and GSK-3β-dependent activation of Wnt/β-catenin signaling pathway in primary rat osteoblasts. Our findings provide novel insights into the mechanisms of action of fluoride in osteoblastogenesis.
Product Description
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Sodium Fluoride Solution