Merck
  • Combination of paclitaxel and bevacizumab in heavily pre-treated non-small-cell lung cancer (NSCLC) patients: a case series study on 15 patients.
Combination of paclitaxel and bevacizumab in heavily pre-treated non-small-cell lung cancer (NSCLC) patients: a case series study on 15 patients.
Bulletin du cancer (2013-12-10)
Sylvestre Le Moulec, Julien Hadoux, Eric Gontier, Cyrus Chargari, Carole Helissey, Virginie Lamand, Rachid Tanz, Françoise Farace, Lionel Vedrine, Gérald Bonardel, Jean-Charles Soria, Benjamin Besse
ABSTRACT
The combination of paclitaxel and bevacizumab was EMA-approved as first-line therapy in metastatic breast cancer. Moreover, in vitro studies showed a potential antiangiogenic synergistic effect of paclitaxel and bevacizumab. Between November 2008 and March 2010, this case series study included 15 patients with metastatic non squamous-cell lung carcinoma (NSCLC). Those were bevacizumab eligible and received the same regimen used in metastatic breast cancer with weekly paclitaxel (80 mg/m(2), days 1, 8 and 15) and bevacizumab (10 mg/kg at days 1 and 15) after at least one prior line of chemotherapy. Efficacy was evaluated by CT-scan and PET-FDG every two months. Circulating endothelial progenitor cells (CEP) and circulating endothelial cells (CEC) levels were explored in a subset of patients. Median age 56 (36-75), female: 47%, never smokers: 27%, adenocarcinoma: 100%, PS 0-1: 87% and PS 3: 13%. All patients were treated with a first-line platinum-based doublet with or without bevacizumab and 70% of them with erlotinib in the second-line. No major toxicity was observed. Partial response (PR) rate was 44% (31-63%) using RECIST criteria on CT-scan, and 65% (29-88%) with PET FDG. PS improved in 33% of the cases. Median progression free survival was 4.6 months. An increase of CEC and CEP was observed in patients with NSCLC treated with paclitaxel and bevacizumab. In this retrospective series, our results suggest efficacy signal in pre-treated metastatic NSCLC and warrant further assessment in a randomized clinical trial.
Product Description
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard