Merck
  • Site-specific aspartic acid isomerization regulates self-assembly and neurotoxicity of amyloid-β.
Site-specific aspartic acid isomerization regulates self-assembly and neurotoxicity of amyloid-β.
Biochemical and biophysical research communications (2014-01-03)
Toshihiko Sugiki, Naoko Utsunomiya-Tate
ABSTRACT
Amyloid-β (Aβ) proteins, which consist of 42 amino acids (Aβ1–42), are the major constituent of neuritic plaques that form in the brains of senile patients with Alzheimer’s disease (AD). Several reports state that three aspartic acid (Asp) residues at positions 1, 7, and 23 in Aβ1–42 in the plaques of patients with AD are highly isomerized from the L- to D-form. Using biophysical experiments, the present study shows that simultaneous D-isomerization of Asp residues at positions 7 and 23 (D-Asp(7,23)) enhances oligomerization, fibril formation, and neurotoxic effect of Aβ1–42. In addition, D-isomerization of Asp at position 1 (D-Asp(1)) suppresses malignant effects induced by D-Asp(7,23) of Aβ1–42. These results provide fundamental information to elucidate molecular mechanisms of AD pathogenesis and to develop potent inhibitors of amyloid aggregates and Aβ neurotoxicity.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)