Merck
  • Military aircrew and noise-induced hearing loss: prevention and management.
Military aircrew and noise-induced hearing loss: prevention and management.
Aviation, space, and environmental medicine (2014-01-28)
Renu Rajguru
ABSTRACT
Modern-day high performance aircraft are more powerful, more efficient, and, unfortunately, frequently produce high noise levels, resulting in noise-induced hearing loss (NIHL) in military aircrew. Military pilots are required to perform many flight duties correctly in the midst of many challenges that may affect mission completion as well as aircraft and aircrew safety. NIHL can interfere with successful mission completion. NIHL may also require aircrew to be downgraded from flying duties, with the incumbent re-training costs for downgraded personnel and training costs for new/replacement aircrew. As it is not possible to control the source of the noise without compromising the efficiency of the engine and aircraft, protecting the aircrew from hazards of excessive noise and treating NIHL are of extreme importance. In this article we discuss various personal hearing protection devices and their efficacy, and pharmacological agents for prevention and management of NIHL.
Product Description
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ebselen, cysteine modifier
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
SAFC
L-Methionine
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Creatine, anhydrous
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Methionine, European Pharmacopoeia (EP) Reference Standard
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%