Merck
  • Synthesis of carboxymethyl chitosan and coating on wound dressing gauze for wound healing.
Synthesis of carboxymethyl chitosan and coating on wound dressing gauze for wound healing.
Pakistan journal of biological sciences : PJBS (2014-02-12)
B Venkatrajah, V Vanitha Malathy, B Elayarajah, R Rajendran, Ram Rammohan
ABSTRACT
Wound healing is a long and complex process. To improve wound healing, the wound dressing cotton gauze can be functionalized by imparting moisture holding and antibacterial ability. Moisture is an important factor for wound healing and the absence of microbial intervention can accelerate wound healing process. Direct alkylation method was used to synthesis carboxymethylated chitosan with water solubility, biocompatibility and antibacterial activity. Calcium alginate was used along with modified chitosan as moisture gaining polymeric agent. Pad-dry-cure method was employed to coat both the polymers on cotton gauze surface, which was weaved using 40s Ne cotton yarn. After coating, the cotton was analysed for its polymer add-on percentage, antibacterial action against Staphylococcus aureus ATCC 6538 and Escherichia coli ATCC 10229. The persistence analysis of antibacterial activity ensures the polymer withstanding ability on cotton gauze surface. SEM detection of polymers with cotton threads confirms their presence. Wound healing action of the polymer coated cotton gauze was determined using albino rats as animal model.
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water
Sigma-Aldrich
Water, HPLC Plus
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Sodium alginate