Merck
  • Magnetic alginate-layered double hydroxide composites for phosphate removal.
Magnetic alginate-layered double hydroxide composites for phosphate removal.
Environmental technology (2014-02-18)
Chang-Gu Lee, Song-Bae Kim
ABSTRACT
The objective of this study was to investigate phosphate removal using magnetic alginate-layered double hydroxide (LDH) composites. The magnetic composites were prepared by entrapping synthetic magnetic iron oxide and calcined Mg-Al LDH in polymer matrix (alginate). Results showed that the magnetic composites (2% magnetic iron oxide and 6% calcined Mg-Al LDH) were effective in the removal of phosphate with the sorption capacity of 5.0 +/- 0.1 mgP/g under given experimental conditions (adsorbent dose = 0.05 g in 30 ml solution; initial phosphate concentration = 10 mgP/l; reaction time = 24 h). Both magnetic iron oxide and calcined Mg-Al LDH have the ability to adsorb phosphate, with the latter having much higher sorption capacity. In the magnetic composites, calcined Mg-Al LDH functions as a phosphate adsorbent while magnetic iron oxide provides both magnetic and sorption properties. Results also demonstrated that phosphate sorption to the magnetic composites reached equilibrium at 24 h. The maximum phosphate sorption capacity was determined to be 39.1 mgP/g. In addition, phosphate removal was not sensitive to initial solution pH between 4.1 and 10.2. Only 9% of the phosphate sorption capacity was reduced as the solution pH increased from 4.1 to 10.2. This study demonstrated that magnetic alginate-LDH composites could be used for phosphate removal in combination with magnetic separation.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Alginic acid from brown algae, powder
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Iron(III) oxide, powder, <5 μm, ≥96%
Sigma-Aldrich
Iron(III) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Iron(III) oxide, nanopowder, <50 nm particle size
Sigma-Aldrich
Magnesium hydroxide, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Aluminum hydroxide, reagent grade
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Water, ACS reagent
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, HPLC Plus
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, Molecular Biology, sterile filtered