Merck
  • Molecular nanofibers of olsalazine form supramolecular hydrogels for reductive release of an anti-inflammatory agent.
Molecular nanofibers of olsalazine form supramolecular hydrogels for reductive release of an anti-inflammatory agent.
Journal of the American Chemical Society (2010-12-03)
Xinming Li, Jiayang Li, Yuan Gao, Yi Kuang, Junfeng Shi, Bing Xu
ABSTRACT
Conjugation of tripeptide derivatives with olsalazine, a clinically used anti-inflammatory prodrug, yields small molecules that self-assemble in water to form supramolecular hydrogels that undergo a gel-to-sol phase transition upon reduction, resulting in the controlled release of 5-aminosalicylic acid as the anti-inflammatory agent. This methodology will ultimately lead to new biomaterials for site-specific drug delivery.
Product Description
Olsalazine sodium for performance test, European Pharmacopoeia (EP) Reference Standard
Olsalazine sodium, European Pharmacopoeia (EP) Reference Standard