Merck
  • Mutagenicity studies with nitrofurans. III. Mutagenicity testing of nitrofurylacrylic acid in human blood and urine.
Mutagenicity studies with nitrofurans. III. Mutagenicity testing of nitrofurylacrylic acid in human blood and urine.
Mutation research (1980-01-01)
M Cerná, P Rössner, K Angelis, J Nováková, R J Srám
ABSTRACT
Human blood and urine mutagenicity of 3-(5-nitro-2-furyl)acrylic acid (5-NFA) was analysed by using Salmonella typhimurium indicator strains TA100 AND TA98 and the cytogenetic analysis of human peripheral lymphocytes. 8 human volunteers were given doses of 1 g 5-NFA per os. The mutagenic effect in blood was analysed after 0, 0.5, 1, 2 and 4 h, in urine after 0, 2 and 4 h. Cytogenetic analysis was done 0, 24 and 72 h after administration of 5-NFA. The experiment was repeated with 3 volunteers in the course of 96 h. When each of 8 volunteers consumed 1 g of 5-NFA, the mutagenicity was observed in 6 blood samples 1 h after exposure for strain TA98 (doubled number or revertants) and in all urine samples taken between the 2nd and 6th hours for both strains used. 7 volunteers given 10 mg 5-NFA in wine (2 sets) showed no mutagenicity of blood or urine for TA100 or TA98 indicator strains. These results are believed to indicate an enhanced elimination of 5-NFA from the human body.