Merck
  • A missense mutation in Rev7 disrupts formation of Polζ, impairing mouse development and repair of genotoxic agent-induced DNA lesions.
A missense mutation in Rev7 disrupts formation of Polζ, impairing mouse development and repair of genotoxic agent-induced DNA lesions.
The Journal of biological chemistry (2013-12-21)
Maryam Khalaj, Abdolrahim Abbasi, Hiroshi Yamanishi, Kouyou Akiyama, Shuso Wakitani, Sotaro Kikuchi, Michiko Hirose, Misako Yuzuriha, Masaki Magari, Heba A Degheidy, Kuniya Abe, Atsuo Ogura, Hiroshi Hashimoto, Tetsuo Kunieda
ABSTRACT
Repro22 is a mutant mouse produced via N-ethyl-N-nitrosourea-induced mutagenesis that shows sterility with germ cell depletion caused by defective proliferation of primordial germ cells, decreased body weight, and partial lethality during embryonic development. Using a positional cloning strategy, we identified a missense mutation in Rev7/Mad2l2 (Rev7(C70R)) and confirmed that the mutation is the cause of the defects in repro22 mice through transgenic rescue with normal Rev7. Rev7/Mad2l2 encodes a subunit of DNA polymerase ζ (Polζ), 1 of 10 translesion DNA synthesis polymerases known in mammals. The mutant REV7 did not interact with REV3, the catalytic subunit of Polζ. Rev7(C70R/C70R) cells showed decreased proliferation, increased apoptosis, and arrest in S phase with extensive γH2AX foci in nuclei that indicated accumulation of DNA damage after treatment with the genotoxic agent mitomycin C. The Rev7(C70R) mutation does not affect the mitotic spindle assembly checkpoint. These results demonstrated that Rev7 is essential in resolving the replication stalls caused by DNA damage during S phase. We concluded that Rev7 is required for primordial germ cell proliferation and embryonic viability and development through the translesion DNA synthesis activity of Polζ preserving DNA integrity during cell proliferation, which is required in highly proliferating embryonic cells.
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2