Merck
  • Antioxidant Vitamin C attenuates experimental abdominal aortic aneurysm development in an elastase-induced rat model.
Antioxidant Vitamin C attenuates experimental abdominal aortic aneurysm development in an elastase-induced rat model.
The Journal of surgical research (2014-02-04)
Tao Shang, Zhao Liu, Chang-jian Liu
ABSTRACT
We investigated the hypothesis that an antioxidant, Vitamin C, could attenuate abdominal aortic aneurysm (AAA) development in a rat model. An AAA model induced by intraluminal infusion was created in 36 male Sprague Dawley rats, which were randomly distributed into three groups: Sham (saline infused, placebo treated), Control (elastase infused, placebo treated), and Vitamin C (elastase infused, vitamin C treated). Vitamin C and placebo were intraperitoneally injected, initiating 1 wk before the infusion and continuing throughout the study. The aortic dilatation ratio was measured, and aortic tissues were further examined using biochemical and histologic techniques. Vitamin C attenuated the development of AAA, decreasing maximal aortic diameter by 25.8% (P < 0.05) and preserving elastin lamellae (P < 0.05). Vitamin C also decreased 8-hydroxyguanine (a marker of oxidative damage to DNA) and 8-isoprostane content (a marker of oxidative stress) in aortic tissues (P < 0.05, respectively). The proteins of matrix metalloproteinase (MMP)-2, MMP-9, and interleukin 6 were markedly downregulated (P < 0.05, respectively), accompanied with notably reduced messenger RNA expression of tumor necrosis factor-α, MMP-2/9, and interleukin 1β (P < 0.05, respectively). However, messenger RNA of tissue inhibitors of metalloproteinase-1 and tissue inhibitors of metalloproteinase-2 were both significantly upregulated in Vitamin C group. Vitamin C treatment had no significant effect on systolic blood pressure (P > 0.05). Vitamin C attenuated AAA development in an elastase-induced rat model via crucial protective effect, which was mediated by an increased level of antioxidant in cooperation with preserving elastin lamellae, inhibiting matrix-degrading proteinases and suppressing inflammatory responses.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Elastase from porcine pancreas, Type I, ≥4.0 units/mg protein
Sigma-Aldrich
Elastase from porcine pancreas, Type III, lyophilized powder, Protein 55-85 %, ≥4.0 units/mg protein
Sigma-Aldrich
Elastase from porcine pancreas, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Elastase from porcine pancreas, Type IV, Protein 50-90 %, lyophilized powder, ≥4.0 units/mg protein (biuret)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard