Merck
  • [Antimicrobial activity of several drugs against extended-spectrum beta-lactamase positive Enterobacteriaceae isolates in Gifu and Aichi prefecture].
[Antimicrobial activity of several drugs against extended-spectrum beta-lactamase positive Enterobacteriaceae isolates in Gifu and Aichi prefecture].
The Japanese journal of antibiotics (2014-02-18)
Satoshi Nakagawa, Harumi Hisada, Nobuhiko Nomura, Junichi Mitsuyama, Shigenori Matsubara, Kazukiyo Yamaoka, Kunitomo Watanabe, Yuko Asano, Hiroyuki Suematsu, Haruki Sawamura, Hikonori Hashido, Yuka Yamagishi, Hiroshige Mikamo, Yoko Matsukawa
ABSTRACT
We investigated the antimicrobial activity of several drugs against 131 extended-spectrum beta-lactamase (ESBL) positive clinical isolates in Gifu and Aichi prefecture from 2007 to 2011. Meropenem (MEPM) and doripenem (DRPM) gave the lowest MIC50 at 0.0313 microg/mL. MEPM gave the lowest MIC90 at 0.0625 microg/mL. According to the Clinical and Laboratory Standards Institute breakpoints, the susceptible rates of carbapenems, tazobactam/piperacillin (TAZ/PIPC) and cefmetazole (CMZ) were higher than 90%. The susceptible rates of MEPM, DRPM, imipenem (IPM), TAZ/PIPC and CMZ were 98.5%, 98.5%, 94.7%, 94.7% and 92.4%. We used the PCR method and identified the molecular types of the ESBL positive isolates. Seventy-two strains had CTX-M-9 group gene and CTX-M-9 group gene is the most frequently detected. Against the CTX-M-9 group gene harboring strains which were the most common in our investigation, the susceptible rates of TAZ/PIPC, MEPM, DRPM and IPM were 100%. It is suggested that not only carbapenems but also TAZ/PIPC and CMZ are useful against infections caused by ESBL positive isolates.
Sigma-Aldrich
β-Lactamase, recombinant, expressed in E. coli
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized, powder, white, ~13 U/mg
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Pseudomonas fluorescens, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, ≥1500.00 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase Blend, recombinant, expressed in E. coli