Merck
  • Monounsaturated fatty acids in serum triacylglycerols are associated with response to neoadjuvant chemotherapy in breast cancer patients.
Monounsaturated fatty acids in serum triacylglycerols are associated with response to neoadjuvant chemotherapy in breast cancer patients.
International journal of cancer (2013-10-12)
Mika Hilvo, Stephan Gade, Tuulia Hyötyläinen, Valentina Nekljudova, Tuulikki Seppänen-Laakso, Marko Sysi-Aho, Michael Untch, Jens Huober, Gunter von Minckwitz, Carsten Denkert, Matej Orešič, Sibylle Loibl
ABSTRACT
Changes in cellular lipid metabolism are a common feature in most solid tumors, which occur already in early stages of the tumor progression. However, it remains unclear if the tumor-specific lipid changes can be detected at the level of systemic lipid metabolism. The objective of this study was to perform comprehensive analysis of lipids in breast cancer patient serum samples. Lipidomic profiling using an established analytical platform was performed in two cohorts of breast cancer patients receiving neoadjuvant chemotherapy. The analyses were performed for 142 patients before and after neoadjuvant chemotherapy, and the results before chemotherapy were validated in an independent cohort of 194 patients. The analyses revealed that in general the tumor characteristics are not reflected in the serum samples. However, there was an association of specific triacylglycerols (TGs) in patients' response to chemotherapy. These TGs containing mainly oleic acid (C18:1) were found in lower levels in those patients showing pathologic complete response before receiving chemotherapy. Some of these TGs were also associated with estrogen receptor status and overall or disease-free survival of the patients. The results suggest that the altered serum levels of oleic acid in breast cancer patients are associated with their response to chemotherapy.
Product Description
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture