Merck
  • Degradation and metabolism of mometasone furoate in humans: influence of reversible, sequential metabolism, and ionic strength.
Degradation and metabolism of mometasone furoate in humans: influence of reversible, sequential metabolism, and ionic strength.
Journal of pharmaceutical sciences (2004-09-29)
Neal M Davies
Product Description
USP
Mometasone furoate, United States Pharmacopeia (USP) Reference Standard
Mometasone furoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mometasone furoate, ≥98% (HPLC)
Supelco
Mometasone Furoate, Pharmaceutical Secondary Standard; Certified Reference Material