Merck
Ciclopirox 1% shampoo for the treatment of seborrheic dermatitis.
International journal of dermatology (2006-01-24)
Aditya K Gupta, Karyn A Nicol
ABSTRACT
Seborrheic dermatitis is a chronic superficial fungal infection of the skin, particularly affecting sites rich in sebaceous glands. Although the precise etiology of seborrheic dermatitis is uncertain, yeasts of the genus Malassezia are known to play a causative role. Ciclopirox is a broad-spectrum, hydroxypyridone-derived, synthetic antifungal agent, which also has anti-inflammatory properties. Ciclopirox is effective both in vitro and in vivo against Malassezia yeasts, making it a valuable option for the treatment of seborrheic dermatitis. Varying frequencies and concentrations of ciclopirox shampoo have been shown to be effective and safe in the treatment of seborrheic dermatitis of the scalp.
Product Description
Ciclopirox, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.