Merck
  • Long-acting opioid-agonists in the treatment of heroin addiction: why should we call them "substitution"?
Long-acting opioid-agonists in the treatment of heroin addiction: why should we call them "substitution"?
Substance use & misuse (2009-04-11)
G Gerra, I Maremmani, B Capovani, L Somaini, S Berterame, J Tomas-Rossello, E Saenz, A Busse, H Kleber
ABSTRACT
Many studies have documented the safety, efficacy, and effectiveness of long-acting opioids (L-AOs), such as methadone and buprenorphine, in the treatment of heroin addiction. This article reviews the pharmacological differences between L-AO medications and short-acting opioids (heroin) in terms of reinforcing properties, pharmacokinetics, effects on the endocrine and immune systems. Given their specific pharmacological profile, L-AOs contribute to control addictive behavior, reduce craving, and restore the balance of disrupted endocrine function. The use of the term "substitution," referring to the fact that methadone or buprenorphine replace heroin in binding to brain opioid receptors, has been generalized to consider L-AOs as simple replacement of street drugs, thus contributing to the widespread misunderstanding of this treatment approach.
Product Description
Supelco
Heroin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Heroin, analytical standard, ≥98% (HPLC)