Merck
  • Combination therapy for managing difficult-to-treat patients with stage 2 hypertension: focus on valsartan-based combinations.
Combination therapy for managing difficult-to-treat patients with stage 2 hypertension: focus on valsartan-based combinations.
American journal of therapeutics (2010-06-11)
Howard S Weintraub, Amy Rudolph
ABSTRACT
Blood pressure (BP) control rates in the United States remain low despite the availability of a wide array of treatment options. High-risk populations, including women, black individuals, and obese patients are even less likely to achieve BP targets than the general population. Uncontrolled BP can lead to serious consequences. Therefore, additional effective and well-tolerated treatment strategies are necessary, particularly for these difficult-to-treat populations. The majority of patients require 2 or more pharmacologic agents to achieve BP control. In patients with stage 2 hypertension, current guidelines recommend rational antihypertensive combinations as initial therapy. The renin-angiotensin-aldosterone system (RAAS) plays an important role in development of hypertension and progression, and the use of RAAS inhibitors has shown BP-lowering effectiveness and excellent tolerability across a continuum of patient types. Agents that block the RAAS form a reasonable foundation for combination therapy. In this article, we review data from studies of valsartan-based antihypertensive combinations in stage 2 hypertension, with a focus on women, black individuals, and obese patients.
Product Description
Valsartan for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Valsartan, United States Pharmacopeia (USP) Reference Standard
Valsartan, European Pharmacopoeia (EP) Reference Standard
Valsartan for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Supelco
Valsartan, Pharmaceutical Secondary Standard; Certified Reference Material