Merck
  • Alopecia areata during treatment of psoriasis with adalimumab and leflunomide: a case and review of the literature.
Alopecia areata during treatment of psoriasis with adalimumab and leflunomide: a case and review of the literature.
Skin pharmacology and physiology (2012-02-04)
R Navarro, E Daudén, E Gallo, D Santiago Sánchez-Mateos, A García-Diez
ABSTRACT
The pathogenesis of alopecia areata (AA) is not clear, even though it is believed to be an autoimmune mechanism which involves T lymphocytes and cytokines such as tumor necrosis factor α. We report the case of a 43-year-old man with psoriasis and psoriatic arthritis who developed AA during his treatment with adalimumab and leflunomide. We perform a review of the literature associating AA with biological therapy and leflunomide. We cannot exclude that the use of these therapies and the development of AA could be coincidental. However, we consider that case reports like ours are essential for clinicians as early alerts if similar observations occur.
Product Description
Leflunomide for peak identification, European Pharmacopoeia (EP) Reference Standard
Leflunomide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Leflunomide, Immunosuppressant
Supelco
Leflunomide, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Leflunomide, United States Pharmacopeia (USP) Reference Standard