Merck
Methotrexate in rheumatoid arthritis: a quarter century of development.
Transactions of the American Clinical and Climatological Association (2013-07-23)
Michael E Weinblatt
ABSTRACT
Methotrexate (MTX) is now the most popular drug worldwide for the treatment of rheumatoid arthritis. Low-dose, weekly MTX (10 to 25 mg/wk) used as either monotherapy or in combination with other drugs has a superior efficacy profile as defined in placebo-controlled trials and comparable efficacy to other drugs including anti-TNF therapy. At 1 year, one third of patients on MTX have no radiographic progression and even greater effects are seen when combined with targeted biological therapies. MTX is well tolerated; gastrointestinal toxicity is the most common toxicity with rarely bone marrow, lung, or liver toxicity. MTX therapy has been a major advance in the treatment of rheumatoid arthritis and is now the cornerstone of therapy.
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)