Merck
  • [Evaluation of the knowledge, attitude and practices of orthodontists in relation to nickel allergy during orthodontic treatment].
[Evaluation of the knowledge, attitude and practices of orthodontists in relation to nickel allergy during orthodontic treatment].
Odonto-stomatologie tropicale = Tropical dental journal (2014-03-15)
W Rerhrhaye, L Bahije, F Zaoui, N Merzouk
ABSTRACT
The aim of this study was to evaluate knowledge, attitudes and practices (KAP) of Moroccan dentists about allergy to Nickel (Ni) during orthodontic treatment. Dentists of two Moroccan cities (n = 553) were contacted and a questionnaire was distributed to those who have certified of partial or exclusive practice of orthodontics (N = 202). They were asked to specify the adverse reactions observed and the alloys frequently used. KAP about etiology, prevention and therapeutic management of allergic patients were also addressed. Twenty-two percent of the dentists have reported at least one allergic patient reaction with more implication of fixed appliances. Eleven percent of adverse effects locations reported were mucosal area and facial skin near contact area and less than 5% was systemic eruption outside contact site. Only 34% of the respondents knew the Nickel allergy etiology. Nickel-containing orthodontic alloys are generally used even in patient with suspected nickel allergy. It is very important to minimize corrosion factors and use nickel-free appliances in clinical practice.
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Nickel, pellets, 50g, max. size 10mm, 99.999%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Nickel, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Nickel, foil, light tested, 100x100mm, thickness 0.025mm, 99.9%
Nickel, wire reel, 5m, diameter 0.025mm, as drawn, 99.98%
Nickel, foil, 5m coil, thickness 0.05mm, annealed, 99%
Nickel, wire reel, 0.1m, diameter 0.75mm, as drawn, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.025mm, 99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Nickel, foil, 0.5m coil, thickness 0.0075mm, 99.95%
Nickel, foil, 0.2m coil, thickness 0.005mm, as rolled, 99.9%
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Nickel, foil, light tested, 100x100mm, thickness 0.05mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.0125mm, 99.9%
Nickel, foil, not light tested, 300x300mm, thickness 0.0125mm, 99.9%
Nickel, foil, not light tested, 25x25mm, thickness 0.001mm, 99.95%
Nickel, foil, 1m coil, thickness 0.075mm, annealed, 99%
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Nickel, mesh, 30x30mm, nominal aperture 0.34mm, thickness 0.025mm, wire diameter 0.041mm, 70 wires/inch, open area 80%, electro-formed mesh, 99.9%
Nickel, rod, 1000mm, diameter 3.0mm, 99%
Nickel, foil, 0.5m coil, thickness 0.125mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.05mm, 99.9%
Nickel, foil, light tested, 300x300mm, thickness 0.02mm, 99.9%
Nickel, foil, light tested, 300x300mm, thickness 0.025mm, 99.9%
Nickel, wire reel, 5m, diameter 2.0mm, hard, 99.98%
Nickel, foil, light tested, 100x100mm, thickness 0.007mm, 99.95%
Nickel, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99%