Merck
  • Absence of high anion gap metabolic acidosis in severe ethylene glycol poisoning: a potential effect of simultaneous lithium carbonate ingestion.
Absence of high anion gap metabolic acidosis in severe ethylene glycol poisoning: a potential effect of simultaneous lithium carbonate ingestion.
American journal of kidney diseases : the official journal of the National Kidney Foundation (1994-02-01)
M Leon, C Graeber
ABSTRACT
Ethylene glycol poisoning is accompanied by a high anion gap metabolic acidosis. We describe a case of severe ethylene glycol poisoning in which severe acidosis was not present. This appears to be due to a beneficial effect exerted by simultaneous ingestion of 80 lithium carbonate tablets, each containing approximately 4 mEq carbonate, with a potential bicarbonate load of 320 mEq. We postulate a protective effect of lithium carbonate due to the bicarbonate generated by this substance.
Sigma-Aldrich
Lithium carbonate, ACS reagent, ≥99.0%
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Sigma-Aldrich
Lithium carbonate, puriss. p.a., ACS reagent, ≥99.0% (T)
Sigma-Aldrich
Lithium carbonate, 99.999% trace metals basis