Merck
  • Mechanisms of lidocaine's action on subtypes of spinal dorsal horn neurons subject to the diverse roles of Na(+) and K(+) channels in action potential generation.
Mechanisms of lidocaine's action on subtypes of spinal dorsal horn neurons subject to the diverse roles of Na(+) and K(+) channels in action potential generation.
Anesthesia and analgesia (2014-06-04)
Matthias Wolff, Rose Schnöbel-Ehehalt, Jörg Mühling, Markus A Weigand, Andrea Olschewski
ABSTRACT
Superficial dorsal horn neurons of the spinal cord receive sensory information from Aδ and C fibers. According to their response to sustained depolarization, these cells can be divided into 3 groups: tonic (TFN), adapting (AFN), and single spike firing (SSN) neurons. During spinal and systemic administration of lidocaine, these neurons are exposed to different concentrations of the local anesthetic lidocaine. In this study, we explored its effect on the excitability of sensory neurons. Whole-cell patch-clamp recordings from dorsal horn neurons of Wistar rats were used to study the action of lidocaine on firing properties. To estimate the impact of a blockade of voltage-gated potassium channels by lidocaine (100 μM) on the firing properties of different neurons, the sodium and potassium channel inhibition of lidocaine was investigated in the light of the effects of tetrodotoxin (TTX, 10 nM) and tetraethylammonium (10 mM). For statistical analysis, the Wilcoxon matched-pairs signed rank test was used throughout. All 3 types of neurons responded to lidocaine with changes in the shape of their action potentials. The peak amplitude of the single action potentials was decreased (P = 0.031, P = 0.013, and P = 0.014 for SSN, AFN, and TFN neurons, respectively), and the duration of the action potentials was increased (P = 0.016, P = 0.032, and P = 0.031 for SSN, AFN, and TFN neurons, respectively). The maximum positive slope (P = 0.016 and P = 0.0010 for SSN and AFN, respectively) and the negative slope (P = 0.016, P = 0.0025, and P = 0.020 for SSN, AFN, and TFN neurons, respectively) decreased after application of lidocaine. In tonically firing neurons, lidocaine reduced the repetitive firing (P = 0.0016), and this effect was mimicked by a combination of TTX and tetraethylammonium. In AFN, TTX mimicked the action of lidocaine. Lidocaine at low concentrations suppresses tonic firing neurons by interacting with voltage-gated potassium channels. The effects on adapting firing neurons can be explained by an interaction with voltage-gated sodium channels. In contrast, the firing pattern of SSN is not affected at the administered concentrations. This different sensitivity to low concentrations of sodium and particularly of potassium channel blockers might represent a novel approach for a differentiated blockade of different spinal dorsal horn neurons.
Product Description
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine hydrochloride monohydrate, solid
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®