Merck
  • Physiology, genetics, and biochemistry of carbon metabolism in the alphaproteobacterium Sinorhizobium meliloti.
Physiology, genetics, and biochemistry of carbon metabolism in the alphaproteobacterium Sinorhizobium meliloti.
Canadian journal of microbiology (2014-08-06)
Barney A Geddes, Ivan J Oresnik
ABSTRACT
A large proportion of genes within a genome encode proteins that play a role in metabolism. The Alphaproteobacteria are a ubiquitous group of bacteria that play a major role in a number of environments. For well over 50 years, carbon metabolism in Rhizobium has been studied at biochemical and genetic levels. Here, we review the pre- and post-genomics literature of the metabolism of the alphaproteobacterium Sinorhizobium meliloti. This review provides an overview of carbon metabolism that is useful to readers interested in this organism and to those working on other organisms that do not follow other model system paradigms.
Product Description
Carbon, foil, 25x25mm, thickness 0.5mm, pyrolytic graphite, 99.99%
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 50mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets