Merck
  • Characterization of a cetyltrimethyl ammonium bromide-modified sorbent for removal of perfluorooctane sulphonate from water.
Characterization of a cetyltrimethyl ammonium bromide-modified sorbent for removal of perfluorooctane sulphonate from water.
Environmental technology (2014-08-26)
Yin-Ming Li, Fu-Shen Zhang
ABSTRACT
This study was carried out to develop a cost-effective and practicable sorbent for application in abrupt perfluorooctane sulphonate (PFOS) pollution accidents. The main merit of this work was that a waste material, namely construction and demolition (C&D) waste, was employed as a raw base material for the sorbent synthesis. The waste material underwent alkaline fusion-hydrothermal synthesis and a cationic surfactant cetyltrimethyl ammonium bromide (CTAB) modification process to form a CTAB-modified sorbent (CMCDSS). Experimental results showed that PFOS concentrations and solution pH had significant effect on the PFOS sorption on construction and demolition waste synthesized sorbent (CDSS) and CMCDSS (using 0.2CMCDSS as representative). PFOS could be effectively and rapidly adsorbed on CMCDSS, and sorption equilibrium was achieved within 2.5 h. The sorption amounts of PFOS on CMCDSSs enhanced along with the increase in CTAB loading amounts. Moreover, the CMCDSS can be applied effectively under acidic condition at pH 2-6 and various removal mechanisms were clarified at different sorption conditions. Accordingly, this work developed a novel and applicable material for dealing with abrupt environmental PFOS contamination accidents.
Product Description
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%