Merck
  • Impact of sodium butyrate on the network of adhesion/growth-regulatory galectins in human colon cancer in vitro.
Impact of sodium butyrate on the network of adhesion/growth-regulatory galectins in human colon cancer in vitro.
Anticancer research (2014-10-03)
Eva-Maria Katzenmaier, Sabine André, Jürgen Kopitz, Hans-Joachim Gabius
ABSTRACT
The physiological compound sodium butyrate can induce differentiation in colon cancer cells in vitro. Due to the role of galectins in growth control we explored its effect on this network beyond galectins-1 and -3, with deliberate consideration of the status of microsatellite stability, for nine cell lines. Microscopical monitoring and measurement of alkaline phosphatase activity ascertained butyrate's impact on cells. Monitoring by reverse transcriptase-polymerase chain reaction (RT-PCR) and western blotting with galectin-type-specific probes characterized galectin expression. Controlled by expectable strong up-regulation of galectin-1 and comparatively small effects on galectin-3 regulation for galectins-4, -7, -8 and -9 were reported with no obvious association to microsatellite stability status. Neoexpression of the GAL-12 gene was observed in eight out of nine tested lines. Butyrate affects the galectin network beyond galectins-1 and -3, warranting further cell biological and histochemical studies.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein