Merck
  • FoxO transcription factors support oxidative stress resistance in human chondrocytes.
FoxO transcription factors support oxidative stress resistance in human chondrocytes.
Arthritis & rheumatology (Hoboken, N.J.) (2014-09-05)
Yukio Akasaki, Oscar Alvarez-Garcia, Masahiko Saito, Beatriz Caramés, Yukihide Iwamoto, Martin K Lotz
ABSTRACT
A major signaling pathway that regulates cellular aging is the insulin/insulin-like growth factor 1 (IGF-1)/phosphatidylinositol 3-kinase (PI3K)/Akt/FoxO transcription factor axis. We previously observed that FoxO transcription factors are dysregulated in aged and OA cartilage. The objective of this study was to investigate the impact of down-regulated FoxO transcription factors on chondrocytes. Small interfering RNAs (siRNAs) targeting FOXO1 (siFOXO1) and FOXO3 (siFOXO3) were transfected into human articular chondrocytes. Cell viability following treatment with the oxidant tert-butyl-hydroperoxide (tBHP) was measured by MTT assay. Caspase 3/7 activation and apoptotic cells were examined. Gene and protein expression of antioxidant proteins and autophagy-related proteins and changes in inflammatory mediators following treatment with interleukin-1β were assessed. Cells transfected with FOXO plasmids were also analyzed. Cell viability was significantly reduced by siFOXO after treatment with tBHP. Apoptosis accompanied by caspase activation was significantly increased in siFOXO-transfected chondrocytes. Knockdown of FOXO1 and FOXO1+3 resulted in significant reductions in levels of glutathione peroxidase 1 (GPX-1), catalase, light chain 3 (LC3), Beclin1, and sirtuin 1 (SIRT-1) proteins following treatment with tBHP. In contrast, the constitutive active form of FOXO3 increased cell viability while inducing GPX-1, Beclin1, and LC3 in response to tBHP. Expression and production of ADAMTS-4 and chemerin were significantly increased in siFOXO-transfected chondrocytes. Reduced expression of FoxO transcription factors in chondrocytes increased susceptibility to cell death induced by oxidative stress. This was associated with reduced levels of antioxidant proteins and autophagy-related proteins. Our data provide evidence for a key role of FoxO transcription factors as regulators of chondrocyte oxidative stress resistance and tissue homeostasis.
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard