Merck
  • Alcohol, one-carbon nutrient intake, and risk of colorectal cancer according to tumor methylation level of IGF2 differentially methylated region.
Alcohol, one-carbon nutrient intake, and risk of colorectal cancer according to tumor methylation level of IGF2 differentially methylated region.
The American journal of clinical nutrition (2014-11-21)
Reiko Nishihara, Molin Wang, Zhi Rong Qian, Yoshifumi Baba, Mai Yamauchi, Kosuke Mima, Yasutaka Sukawa, Sun A Kim, Kentaro Inamura, Xuehong Zhang, Kana Wu, Edward L Giovannucci, Andrew T Chan, Charles S Fuchs, Shuji Ogino, Eva S Schernhammer
ABSTRACT
Although a higher consumption of alcohol, which is a methyl-group antagonist, was previously associated with colorectal cancer risk, mechanisms remain poorly understood. We hypothesized that excess alcohol consumption might increase risk of colorectal carcinoma with hypomethylation of insulin-like growth factor 2 (IGF2) differentially methylated region-0 (DMR0), which was previously associated with a worse prognosis. With the use of a molecular pathologic epidemiology database in 2 prospective cohort studies, the Nurses' Health Study and Health Professionals Follow-up Study, we examined the association between alcohol intake and incident colorectal cancer according to the tumor methylation level of IGF2 DMR0. Duplication-method Cox proportional cause-specific hazards regression for competing risk data were used to compute HRs and 95% CIs. In addition, we investigated intakes of vitamin B-6, vitamin B-12, methionine, and folate as exposures. During 3,206,985 person-years of follow-up, we identified 993 rectal and colon cancer cases with an available tumor DNA methylation status. Compared with no alcohol consumption, the consumption of ≥15 g alcohol/d was associated with elevated risk of colorectal cancer with lower levels of IGF2 DMR0 methylation [within the first and second quartiles: HRs of 1.55 (95% CI: 1.08, 2.24) and 2.11 (95% CI: 1.44, 3.07), respectively]. By contrast, alcohol consumption was not associated with cancer with higher levels of IGF2 DMR0 methylation. The association between alcohol and cancer risk differed significantly by IGF2 DMR0 methylation level (P-heterogeneity = 0.006). The association of vitamin B-6, vitamin B-12, and folate intakes with cancer risk did not significantly differ according to IGF2 DMR0 methylation level (P-heterogeneity > 0.2). Higher alcohol consumption was associated with risk of colorectal cancer with IGF2 DMR0 hypomethylation but not risk of cancer with high-level IGF2 DMR0 methylation. The association between alcohol intake and colorectal cancer risk may differ by tumor epigenetic features.
Product Description
Folic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
SAFC
L-Methionine
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethyl alcohol, denatured, denatured, reagent grade
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Methionine, European Pharmacopoeia (EP) Reference Standard