Merck
  • Improved synthesis of 5-substituted 1H-tetrazoles via the [3+2] cycloaddition of nitriles and sodium azide catalyzed by silica sulfuric acid.
Improved synthesis of 5-substituted 1H-tetrazoles via the [3+2] cycloaddition of nitriles and sodium azide catalyzed by silica sulfuric acid.
International journal of molecular sciences (2012-05-19)
Zhenting Du, Changmei Si, Youqiang Li, Yin Wang, Jing Lu
ABSTRACT
A silica supported sulfuric acid catalyzed [3+2] cycloaddition of nitriles and sodium azide to form 5-substituted 1H-tetrazoles is described. The protocol can provide a series of 5-substituted 1H-tetrazoles using silica sulfuric acid from nitriles and sodium azide in DMF in 72%-95% yield.