Merck
  • Degradation of atrazine by microwave-assisted electrodeless discharge mercury lamp in aqueous solution.
Degradation of atrazine by microwave-assisted electrodeless discharge mercury lamp in aqueous solution.
Journal of hazardous materials (2006-07-11)
Na Ta, Jun Hong, Tingfeng Liu, Cheng Sun
ABSTRACT
The present study investigates the degradation of atrazine (2-chloro-4-(ethyl amino)-6-isopropyl amino-s-triazine) in aqueous solution by a developed new method, namely by means of a microwave-assisted electrodeless discharge mercury lamp (MW-EDML). An experimental design was conducted to assess the influence of various parameters: pH value, initial concentration, amount of EDML, initial volume and coexisted solvent. Atrazine was degraded completely by EDML in a relatively short time (i.e. t(1/2)=1.2 min for 10 mg/l). Additionally, the identification of main degradation products during atrazine degradation process was conducted by gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-mass spectrometry (LC-MS). This study proposes the degradation mechanism including four possible pathways for atrazine degradation according to the degradation products.
Product Description
Supelco
Supelclean ENVI-18 SPE Tube, bed wt. 500 mg, volume 6 mL, pkg of 30 ea
Pricing and availability is not currently available.