Merck
  • Detection of microRNA in tumor cells using exonuclease III and graphene oxide-regulated signal amplification.
Detection of microRNA in tumor cells using exonuclease III and graphene oxide-regulated signal amplification.
ACS applied materials & interfaces (2014-04-16)
Rong-Cing Huang, Wei-Jane Chiu, Yu-Jia Li, Chih-Ching Huang
ABSTRACT
In this study, we developed a label-free, ultrasensitive graphene oxide (GO)-based probe for the detection of oligonucleotides by laser desorption/ionization mass spectrometry (LDI-MS). On the basis of simple π-π stacking and electrostatic interactions between rhodamine 6G (R6G) and GO, we prepared the nanocomposite R6G-modified GO (R6G-GO). Signal intensities of R6G increased in mass spectra in the presence of single-stranded oligonucleotides under pulsed laser irradiation (355 nm) of R6G-GO. In addition, the signal intensity of R6G was stronger in the presence of short oligonucleotides. Because small oligonucleotides improve the LDI efficiency of R6G on GO, we designed an enzyme-amplified signal transduction probe system for the detection of microRNA (miRNA). After specific digestion of the probe DNA (pDNA) strand from pDNA/miRNA-hybridized complexes by exonuclease III (Exo III), the resulting small oligonucleotide fragments increased the R6G signal during LDI-MS of R6G-GO. In addition, the signal intensity of the R6G ions increased with increasing concentrations of the target miRNA. Coupling this enzyme reaction and R6G-GO with LDI-MS enabled the detection of miRNA at concentrations of the femtomolar (fM) level. We also demonstrated the analysis of miRNA in tumor cells and utilized this R6G-GO probe in the detection of a single-nucleotide polymorphism (SNP) in the Arg249Ser unit of the TP53 gene. This simple, rapid, and sensitive detection system based on the coupling of functional GO with LDI-MS appears to have great potential as a tool for the bioanalyses of oligonucleotides and proteins.
Product Description
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%