Merck
  • Fluorescent sensors for the basic metabolic panel enable measurement with a smart phone device over the physiological range.
Fluorescent sensors for the basic metabolic panel enable measurement with a smart phone device over the physiological range.
The Analyst (2014-08-16)
Becker Awqatty, Shayak Samaddar, Kevin J Cash, Heather A Clark, J Matthew Dubach
ABSTRACT
The advanced functionality of portable devices such as smart phones provides the necessary hardware to potentially perform complex diagnostic measurements in any setting. Recent research and development have utilized cameras and data acquisition properties of smart phones to create diagnostic approaches for a variety of diseases or pollutants. However, in concentration measurements, such as blood glucose, the performance of handheld diagnostic devices depends largely on the sensing mechanism. To expand measurements to multiple components, often necessary in medical tests, with a single diagnostic device, robust platform based sensors are needed. Here, we developed a suite of dual wavelength fluorescent sensors with response characteristics necessary to measure each component of a basic metabolic panel, a common clinical measurement. Furthermore, the response of these sensors could be measured with a simple optical setup to convert a smart phone into a fluorescence measurement instrument. This approach could be used as a mobile basic metabolic panel measurement system for point of care diagnostics.
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
1-Chlorooctadecane, 96%