Merck
  • Lipid based nanocarrier system for the potential oral delivery of decitabine: formulation design, characterization, ex vivo, and in vivo assessment.
Lipid based nanocarrier system for the potential oral delivery of decitabine: formulation design, characterization, ex vivo, and in vivo assessment.
International journal of pharmaceutics (2014-12-03)
Yub Raj Neupane, Manish Srivastava, Nafees Ahmad, Neeraj Kumar, Aseem Bhatnagar, Kanchan Kohli
ABSTRACT
The aim of this study was to design and fabricate nanostructured lipid carrier (NLC) for the potential oral delivery of decitabine (DCB). NLC was prepared by cold homogenization technique and optimized by the Box-Behnken experimental design. It was further characterized by particle size, zeta potential, transmission electron microscopy (TEM), atomic force microscopy (AFM), differential scanning calorimetry (DSC), X-ray diffraction (XRD), in vitro release study, and stability study. Moreover, ex vivo and in vivo efficacy of the NLC was assessed by gut permeation study, γ scintigraphy imaging, and MTT assay. NLC was found to have particle size (116.64 ± 6.67 nm), zeta potential (-31.8 ± 0.96 mV) and sustained drug release (80.23 ± 4.67%) up to 24h. TEM and AFM proved that the particles were spherical in shape and smooth surface. DSC and XRD studies had demonstrated the reduced crystallinity and stability enhancing effect of the NLC. Stability studies revealed the changes in the observed parameters up to 45 days were not significantly differences (p>0.05). Ex vivo gut permeation study showed 4-folds increment in the permeation of drug compared with the plain drug solution. γ Scintigraphy imaging and MTT assay results inferred that DCB loaded NLC possesses excellent cytotoxic activity against cancer cells. Thus, NLC holds high potential for the oral delivery of DCB to treat cancer cells and future prospects for the industrial purpose.
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)
Supelco
Span® 80, suitable for GC
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%