Merck
  • Evaluation of Hs-CRP levels and interleukin 18 (-137G/C) promoter polymorphism in risk prediction of coronary artery disease in first degree relatives.
Evaluation of Hs-CRP levels and interleukin 18 (-137G/C) promoter polymorphism in risk prediction of coronary artery disease in first degree relatives.
PloS one (2015-03-31)
Rajesh Kumar G, Mrudula Spurthi K, Kishore Kumar G, Mohanalatha Kurapati, Saraswati M, Mohini Aiyengar T, Chiranjeevi P, Srilatha Reddy G, Nivas S, Kaushik P, Sanjib Sahu K, Surekha Rani H
ABSTRACT
Coronary Artery Disease (CAD) is clearly a multifactorial disease that develops from childhood and ultimately leads to death. Several reports revealed having a First Degree Relatives (FDRS) with premature CAD is a significant autonomous risk factor for CAD development. C - reactive protein (CRP) is a member of the pentraxin family and is the most widely studied proinflammatory biomarker. IL-18 is a pleiotrophic and proinflammatory cytokine which is produced mainly by macrophages and plays an important role in the inflammatory cascade. Hs-CRP levels were estimated by ELISA and Genotyping of IL-18 gene variant located on promoter -137 (G/C) by Allele specific PCR in blood samples of 300 CAD patients and 300 controls and 100 FDRS. Promoter Binding sites and Protein interacting partners were identified by Alibaba 2.1 and Genemania online tools respectively. Hs-CRP levels were significantly high in CAD patients followed by FDRS when compared to controls. In IL-18 -137 (G/C) polymorphism homozygous GG is significantly associated with occurrence of CAD and Hs-CRP levels were significantly higher in GG genotype subjects when compared to GC and CC. IL-18 was found to be interacting with 100 protein interactants. Our results indicate that Hs-CRP levels and IL-18-137(G/C) polymorphism may help to identify risk of future events of CAD in asymptomatic healthy FDRS.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O