Merck
  • Biochemical and histopathological evidence on the beneficial effects of Tragopogon graminifolius in TNBS-induced colitis.
Biochemical and histopathological evidence on the beneficial effects of Tragopogon graminifolius in TNBS-induced colitis.
Pharmaceutical biology (2014-12-05)
Mohammad Hosein Farzaei, Seyedeh Farnaz Ghasemi-Niri, Amir Hosein Abdolghafari, Maryam Baeeri, Mahnaz Khanavi, Mona Navaei-Nigjeh, Mohammad Abdollahi, Roja Rahimi
ABSTRACT
Tragopogon graminifolius DC. (Compositae) (TG) has been proposed as an efficacious remedy for gastrointestinal ulcers in Iranian traditional medicine. The present study evaluates the efficacy of TG on experimental colitis and the responsible mechanisms. After induction of IBD by 2,4,6-trinitrobenzenesulfonic acid (TNBS), rats received standardized ethanol extract of TG aerial part at 20, 30, or 50 mg/kg/d orally. After 12 d, the rats were sacrificed and the colon was removed and assessed for macroscopic and microscopic changes. Also, tumor necrosis factor-alpha (TNF-α), interleukin-1 beta (IL-1β), total antioxidant capacity, myeloperoxidase (MPO), and lipid peroxidation (LPO) were measured in the colon homogenate. TG extract significantly reduced macroscopic and microscopic scores of colitis with ED50 values of 23 and 39 mg/kg, respectively. MPO was significantly reduced in all plant extract groups with an ED50 value of 41 mg/kg. The ED50 values of extract for inhibition of TNF-α and LPO were 44 and 93 mg/kg, respectively. IL-1β significantly decreased by 50 mg/kg of TG extract (ED50 = 57 mg/kg). Total antioxidant power markedly increased by 50 mg/kg group (ED50 = 43 mg/kg). TG exhibited efficacy on TNBS-induced colitis via anti-inflammatory, immunomodulatory, antioxidant, and mucosal healing properties. TG possesses promising healing function on colitis. Clinical trials are warranted to prove its efficacy and tolerability in IBD.
Product Description
Supelco
Ethyl acetate, analytical standard
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Picrylsulfonic acid solution, 1 M in H2O
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur