Merck
  • Mutation spectrum of RB1 gene in unilateral retinoblastoma cases from Tunisia and correlations with clinical features.
Mutation spectrum of RB1 gene in unilateral retinoblastoma cases from Tunisia and correlations with clinical features.
PloS one (2015-01-21)
Hajer Ayari-Jeridi, Kimberly Moran, Amel Chebbi, Hédi Bouguila, Imen Abbes, Khaoula Charradi, Amel Benammar-Elgaaïed, Arupa Ganguly
ABSTRACT
Retinoblastoma, an embryonic neoplasm of retinal origin, is the most common primary intraocular malignancy in children. Somatic inactivation of both alleles of the RB1 tumor suppressor gene in a retinal progenitor cell through diverse mechanisms including genetic and epigenetic modifications, is the crucial event in initiation of tumorigenesis in most cases of isolated unilateral retinoblastoma. We analyzed DNA from tumor tissue and from peripheral blood to determine the RB1 mutation status and seek correlations with clinical features of 37 unrelated cases of Tunisian origin with sporadic retinoblastoma. All cases were unilateral except one who presented with bilateral disease, in whom no germline coding sequence alteration was identified. A multi-step mutation scanning protocol identified bi-allelic inactivation of RB1 gene in 30 (81%) of the samples tested. A total of 7 novel mutations were identified. There were three tumors without any detectable mutation while a subset contained multiple mutations in RB1 gene. The latter group included tumors collected after treatment with chemotherapy. There were seven individuals with germline mutations and all presented with advanced stage of tumor. There was no difference in age of onset of RB based on the germline mutation status. Thus 20% of the individuals with sporadic unilateral RB in this series carried germline mutations and indicate the importance of genetic testing all children with sporadic retinoblastoma. These findings help to characterize the spectrum of mutations present in the Tunisian population and can improve genetic diagnosis of retinoblastoma.
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., ACS reagent, ≥99.8% (chloroform + ethanol, GC)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Phenol, PESTANAL®, analytical standard