Merck
  • Potential predictive plasma biomarkers for cervical cancer by 2D-DIGE proteomics and Ingenuity Pathway Analysis.
Potential predictive plasma biomarkers for cervical cancer by 2D-DIGE proteomics and Ingenuity Pathway Analysis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-11-28)
Xia Guo, Yi Hao, Mayila Kamilijiang, Axiangu Hasimu, Jianlin Yuan, Guizhen Wu, Halidan Reyimu, Nafeisha Kadeer, Abulizi Abudula
ABSTRACT
The current methods available for screening and detecting cervical squamous cell carcinoma (CSCC) have insufficient sensitivity and specificity. As a result, many patients suffered from erroneous and missed diagnosis. Because CSCC is usually asymptomatic at potentially curative stages, identification of biomarkers is an urgent need for the early detection of CSCC. Comparative proteomics based on two-dimensional differential in-gel electrophoresis (2D-DIGE) was employed to quantitatively analyze plasma proteins of healthy Uyghur women and with early stage cervical carcinoma. The 2D-DIGE image were analyzed statistically using DeCyder™ 2D software. The statistical analysis of proteomic data revealed that 43 protein spots showed significantly different expression (ratio > 1.5, P < 0.01). A further identification of these protein spots by MALDI-TOF-MS found out 16 different proteins. Bioinformatic analysis within the framework of Ingenuity Pathway Analysis (IPA(@)) showed that 10 plasma proteins as candidate biomarker were screened, mainly including lipid metabolism-related proteins (APOA4, APOA1, APOE), complement (EPPK1, CFHR1), metabolic enzymes (CP, F2, MASP2), glycoprotein (CLU), and immune function-related proteins (IGK@). Networks involved in lipid metabolism, molecular transport, and small molecule biochemistry were dysfunctional in CSCC. Acute phase response signaling and JAK/Stat signaling and IL-4 signaling, etc., were identified as the canonical pathways that are overrepresented in CSCC. Furthermore, the expression of three proteins (APOA1, APOE, CLU) were validated using ELISA in plasma of patients with different stage cervical lesion. With the combined proteomic and bioinformatic approach, this study was successful in identifying biomarker signatures for cervical cancer and might provide new insights into the mechanism of CSCC progression, potentially leading to the design of novel diagnostic and therapeutic strategies.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Glycine, 99%, FCC
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
3-Indoleacetic acid, 98%