Merck
Effects of quetiapine on DNA methylation in neuroblastoma cells.
Progress in neuro-psychopharmacology & biological psychiatry (2014-09-10)
Hiroko Sugawara, Miki Bundo, Tatsuro Asai, Fumiko Sunaga, Junko Ueda, Jun Ishigooka, Kiyoto Kasai, Tadafumi Kato, Kazuya Iwamoto
ABSTRACT
Epigenetic regulation may be involved in the pathophysiology of mental disorders, such as schizophrenia and bipolar disorder, and in the pharmacological action of drugs. Characterizing the epigenetic effects of drugs is an important step to optimal treatment. We performed comprehensive and gene-specific DNA methylation analyses of quetiapine using human neuroblastoma cells. Human neuroblastoma cells were cultured with quetiapine for 8 days, and DNA methylation analysis was performed using Infinium HumanMethylation27 BeadChip. A total of 1173 genes showed altered DNA methylation. Altered DNA methylation predominantly occurred as hypomethylation within the CpG island compared to DNA isolated from non-treated cells. Gene ontology analysis revealed that these genes were related to the cellular process of intracellular protein binding. There was no common effect of quetiapine with three mood stabilizers (lithium, valproate, and carbamazepine). However, common DNA methylation changes in eight genes, including ADRA1A, which encodes adrenoceptor alpha 1A, were found with quetiapine and lithium treatments. Finally, bisulfite-sequencing analysis revealed that quetiapine decreased the DNA methylation level of the promoter region of SLC6A4, where hypermethylation with bipolar disorder and hypomethylation with mood stabilizers have been reported.
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%