Merck
  • Multiresidue determination of ten nonsteroidal anti-inflammatory drugs in bovine, porcine, and chicken liver tissues by HPLC-MS/MS.
Multiresidue determination of ten nonsteroidal anti-inflammatory drugs in bovine, porcine, and chicken liver tissues by HPLC-MS/MS.
Applied biochemistry and biotechnology (2014-07-30)
JeongWoo Kang, Su-Jeong Park, Hae-Chul Park, Vinayakumar Gedi, ByungJae So, Kwang-Jick Lee
ABSTRACT
Nonsteroidal anti-inflammatory drugs (NSAIDs) are the group of drugs having the therapeutic efficacy of analgesic and antipyretic. To detect health-threatening residues of NSAIDs, a fast and easy multiresidue method based on liquid chromatography tandem mass spectrometry (LC-MS/MS) was described. Ten NSAIDs were extracted from the tissues using 2 mL of acetonitrile and 0.1 mL of 2 mM ammonium formate in distilled water. After clean-up using C18 sorbent, it was evaporated under nitrogen, reconstituted with 1 mL distilled water and analyzed by LC-MS/MS. The method was validated based on guideline for residue testing laboratory. Furthermore, the method has also been applied successfully to detect ten NSAIDs from bovine, porcine, and chicken liver tissues. In a total of 315 liver samples tested, acetylic salicylic acid was detected from 28 porcine and 2 chicken liver tissues at levels of 13 ∼ 576 and 50 ∼ 53 ng/g, respectively. Subsequently, paracetamol was detected in 15 porcine liver tissues with a detection levels of 28 ∼ 381 ng/g. Phenylbutazone and its metabolite, oxyphenylbutazone, were detected at 247 and 15 ng/g range in one of the bovine liver tissue, respectively.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Carprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
Carprofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Carprofen, VETRANAL®, analytical standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tolfenamic acid, NSAID
Supelco
Tolfenamic acid, VETRANAL®, analytical standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Supelco
Ketoprofen, VETRANAL®, analytical standard
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Tolfenamic acid, European Pharmacopoeia (EP) Reference Standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Phenylbutazone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Supelco
Phenylbutazone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Phenylbutazone
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Carprofen for system suitability, European Pharmacopoeia (EP) Reference Standard